Merck
  • Enhancement of antiproliferative activity by molecular simplification of catalpol.
Enhancement of antiproliferative activity by molecular simplification of catalpol.
Bioorganic & medicinal chemistry (2010-03-17)
Celina García, Leticia G León, Carlos R Pungitore, Carla Ríos-Luci, Antonio H Daranas, Juan C Montero, Atanasio Pandiella, Carlos E Tonn, Víctor S Martín, José M Padrón
ABSTRACT
Two iridoid scaffolds were synthesized enantioselectively using as key step an l-proline-catalyzed alpha-formyl oxidation. The in vitro antiproliferative activities were evaluated against a representative panel of human solid tumor cell lines. Both iridoids induced considerably growth inhibition in the range 0.38-1.86muM. Cell cycle studies for these compounds showed the induction of cell cycle arrest at the G(1) phase. This result was consistent with a decrease in the expression of cyclin D1. Damaged cells underwent apoptosis as indicated by specific Annexin V staining.
Sigma-Aldrich
Catalpol, ≥96% (HPLC)
Pricing and availability is not currently available.