Merck
  • Inhibition of hypoxia inducible factor 1-transcription coactivator interaction by a hydrogen bond surrogate alpha-helix.
Inhibition of hypoxia inducible factor 1-transcription coactivator interaction by a hydrogen bond surrogate alpha-helix.
Journal of the American Chemical Society (2010-01-01)
Laura K Henchey, Swati Kushal, Ramin Dubey, Ross N Chapman, Bogdan Z Olenyuk, Paramjit S Arora
ABSTRACT
Designed ligands that inhibit hypoxia-inducible gene expression could offer new tools for genomic research and, potentially, drug discovery efforts for the treatment of neovascularization in cancers. We report a stabilized alpha-helix designed to target the binding interface between the C-terminal transactivation domain (C-TAD) of hypoxia-inducible factor 1alpha (HIF-1alpha) and cysteine-histidine rich region (CH1) of transcriptional coactivator CBP/p300. The synthetic helix disrupts the structure and function of this complex, resulting in a rapid downregulation of two hypoxia-inducible genes (VEGF and GLUT1) in cell culture.
Sigma-Aldrich
Chetomin, from Chaetomium cochliodes, ≥98% (HPLC)