Merck
  • New radiochromic gel for 3D dosimetry based on Turnbull blue: basic properties.
New radiochromic gel for 3D dosimetry based on Turnbull blue: basic properties.
Physics in medicine and biology (2009-08-05)
Jaroslav Solc, Václav Spevácek
ABSTRACT
The recently developed new radiochromic gel dosimeter based on Turnbull blue dye formed by irradiation (the TBG dosimeter) does not exhibit dose pattern degradation due to diffusion effects as observed in the Fricke-gel dosimeter with xylenol orange incorporated into the gel matrix (the FXG dosimeter). The TBG dosimeter can be easily prepared and its optical properties enable evaluation of the gel's response using the cone-beam optical computed tomography technique. The preparation procedure is described in the paper along with the basic characteristics of the gel, including dose response, dose sensitivity, ageing under different storage conditions, diffusion rates of Turnbull blue and gel density. The measurement of diffusion is described in more detail. The same method was applied to the FXG dosimeter for direct comparison. It was found that the diffusion coefficient of the TBG dosimeter stored at 24 degrees C is less than 4 x 10(-3) mm(2) h(-1) (1sigma confidence level), compared to the value of 7.3 x 10(-1) mm(2) h(-1) (1sigma) of the FXG dosimeter measured at the same temperature. Although the TBG dosimeter is less sensitive than the FXG dosimeter, its diffusion coefficient is practically negligible and, therefore, it offers large potential as a three-dimensional dosimeter for applications encompassing sharp dose gradients such as high-dose-rate brachytherapy.
Product Description
Supelco
Xylenol Orange disodium salt, for spectrophotometric determination of metal ions
Pricing and availability is not currently available.