Merck
  • Stereoselective total synthesis of cis- and trans-3-hydroxypipecolic acid.
Stereoselective total synthesis of cis- and trans-3-hydroxypipecolic acid.
The Journal of organic chemistry (2005-11-19)
Ningning Liang, Apurba Datta
ABSTRACT
[reaction: see text] 3-Hydroxypipecolic acid, a nonproteinogenic cyclic alpha-amino acid, is a common structural moiety found in a large number of natural and synthetic compounds of medicinal significance. Utilizing D-serine as a chiral template, the present research describes efficient and straightforward routes to cis- and trans-3-hydroxypipecolic acids in enantiopure form. The key steps in the syntheses involve chelation-controlled addition of a homoallyl Grignard reagent to a protected serinal derivative toward stereoselective formation of the corresponding syn-amino alcohol adduct 3. On the other hand, zinc borohydride-mediated chelation-controlled reduction of a serine-derived alpha-aminoketone precursor leads to the formation of the corresponding anti-amino alcohol adduct 4 with high stereoselectivity. Following an efficient sequence of reactions, the above amino alcohol derivatives were subsequently transformed to the corresponding cis- and trans- title compounds, respectively.
Product Description
Supelco
3-Hydroxypicolinic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
3-Hydroxypyridine-2-carboxylic acid, 98%