Merck
  • Preparation and biological properties of dichloro(1,2-diaminocyclohexane)platinum(II) (DACHPt)-loaded polymeric micelles.
Preparation and biological properties of dichloro(1,2-diaminocyclohexane)platinum(II) (DACHPt)-loaded polymeric micelles.
Journal of controlled release : official journal of the Controlled Release Society (2004-12-14)
Horacio Cabral, Nobuhiro Nishiyama, Soichiro Okazaki, Hiroyuki Koyama, Kazunori Kataoka
ABSTRACT
Block copolymer micelles, containing dichloro(1,2-diaminocyclohexane)platinum(II) (DACHPt), the oxaliplatin parent complex, were prepared through polymer-metal complex formation of DACHPt with poly(ethylene glycol)-poly(glutamic acid) block copolymer [PEG-P(Glu)] in distilled water. By dynamic light scattering (DLS) measurement, the micelle size was determined to be 40 nm with narrow distribution. The release of platinum complexes from the micelle core was measured in phosphate buffer saline (pH 7.4) at 37 degrees C. DACHPt-loaded micelle showed a sustained release rate of platinum after an induction period of 12 h. In the same conditions, the kinetic stability of DACHPt-loaded micelle was measured. The micelle was found to be very stable, keeping the initial size, for 240 h. Against murine colon adenocarcinoma 26 (C-26) cells, DACHPt-loaded micelle exhibited considerable in vitro cytotoxicity, lower than oxaliplatin but increasing with exposure time as a result of the release of platinum complexes from the micelle. In vivo biodistribution assay performed on tumor-bearing mice demonstrated that the micelle showed prolonged blood circulation due to its high stability and high tumor accumulation for a prolonged time.
Sigma-Aldrich
Dichloro(1,2-diaminocyclohexane)platinum(II)