Merck
  • New intramolecularly quenched fluorogenic peptide substrates for the study of the kinetic specificity of papain.
New intramolecularly quenched fluorogenic peptide substrates for the study of the kinetic specificity of papain.
FEBS letters (1992-02-03)
C García-Echeverría, D H Rich
ABSTRACT
A series of new substrates for determining the catalytic activity of cysteine proteinases is described. The rate of hydrolysis by papain was monitored by a fluorescence continuous assay based on internal resonance energy transfer using 5-[(2-aminoethyl)amino]naphtalene-1-sulfonic acid (EDANS) and 4-(4-dimethylaminophenylazo)benzoic acid (DABCYL) as fluorescent donor and quenching acceptor, respectively, in peptides with the general structure: DABCYL-Lys-Phe-Gly-Xxx-Ala-Ala-EDANS. The substrates were used to evaluate the effect of amino acid structure in the S1' position on the kinetic parameters for papain catalyzed hydrolysis.
Sigma-Aldrich
4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester, ≥98.0% (HPLC)
Pricing and availability is not currently available.