Merck
  • Tetragonal deformation of the hexagonal myofilament matrix in single skinned skeletal muscle fibres owing to change in sarcomere length.
Tetragonal deformation of the hexagonal myofilament matrix in single skinned skeletal muscle fibres owing to change in sarcomere length.
Journal of muscle research and cell motility (1992-10-01)
P Schiereck, E L de Beer, R L Grundeman, T Manussen, N Kylstra, W Bras
ABSTRACT
Single skinned skeletal muscle fibres were immersed in solutions containing two different levels of activator calcium (pCa: 4.4; 6.0). Sarcomere length was varied from 1.6 to 3.5 microns and recorded by laser diffraction. Slack length was 2.0 microns. Small-angle equatorial X-ray diffraction patterns of relaxed and activated fibres at different sarcomere lengths were recorded using synchrotron radiation. The position and amplitude of the diffraction peaks were calculated from the spectra based on the hexagonal arrangement of the myofilament matrix, relating the position of the (1.0)- and (1.1)-diffraction peaks in this model by square root of 3. The diffraction peaks were fitted by five Gaussian functions (1.0, 1.1, 2.0, 2.1 and Z-line) and residual background was corrected by means of a hyperbola. The coupling of the position of the (1.0)- and (1.1)-peak was expressed as a factor: FAC = [d(1.0)/d(1.1)]/square root 3. In the relaxed state this coupling factor decreased at increasing sarcomere length (0.9880 +/- 0.002 at 2.0 microns; 0.900 +/- 0.01 at 3.5 microns). The coupling factor tends toward the one that will be obtained from the squared structure of actin filaments near the Z-discs. At shorter sarcomere lengths a decrease of the coupling factor has also been seen (0.9600 +/- 0.005 at 1.6 microns), giving rise to an increased uniform deformation of the hexagonal matrix, when sarcomere length is changed from slack length. From these experiments we conclude that a change in sarcomere length (from slack length) increases the deformation of the actin-myosin matrix to a tetragonal lattice.
Sigma-Aldrich
1,6-Diaminohexane-N,N,N′,N′-tetraacetic acid, 97%
Pricing and availability is not currently available.