Merck
  • Long-range structural preformation in yes-associated protein precedes encounter complex formation with TEAD.
Long-range structural preformation in yes-associated protein precedes encounter complex formation with TEAD.
iScience (2022-04-06)
Michael Feichtinger, Andreas Beier, Mario Migotti, Matthias Schmid, Fedir Bokhovchuk, Patrick Chène, Robert Konrat
ABSTRACT
Yes-associated protein (YAP) is a partly intrinsically disordered protein (IDP) that plays a major role as the downstream element of the Hippo pathway. Although the structures of the complex between TEA domain transcription factors (TEADs) and the TEAD-binding domain of YAP are already well characterized, its apo state and the binding mechanism with TEADs are still not clearly defined. Here we characterize via a combination of different NMR approaches with site-directed mutagenesis and affinity measurements the intrinsically disordered solution state of apo YAP. Our results provide evidence that the apo state of YAP adopts several compact conformations that may facilitate the formation of the YAP:TEAD complex. The interplay between local secondary structure element preformation and long-range co-stabilization of these structured elements precedes the encounter complex formation with TEAD and we, therefore, propose that TEAD binding proceeds largely via conformational selection of the preformed compact substates displaying at least nanosecond lifetimes.
Sigma-Aldrich
Tuner (DE3)pLacI Competent Cells - Novagen®, Tuner host strains are lacZY deletion mutants of BL21, which enable adjustable levels of protein expression throughout all cells in a culture. Contains the pLacI plasmid producing extra Lac repressor.
Pricing and availability is not currently available.