Merck
  • NADPH and xanthine oxidases control induction of inflammatory mediator expression by organic dust in the lung.
NADPH and xanthine oxidases control induction of inflammatory mediator expression by organic dust in the lung.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2022-06-07)
Velmurugan Meganathan, Cory E Hamilton, Kartiga Natarajan, Shiva Keshava, Vijay Boggaram
ABSTRACT
Exposure to organic dust in animal and agricultural farms and the ensuing lung inflammation are linked to the development of respiratory diseases. We found previously that elevated production of reactive oxygen species (ROS) by aqueous poultry organic dust extract (hereafter referred to as dust extract) mediates induction of proinflammatory mediators in airway epithelial cells. In the present study, we investigated whether ROS generated by NADPH oxidases (NOX) and xanthine oxidase (XO) controls induction of inflammatory mediators by dust extract and the underlying mechanisms in bronchial epithelial cells. Using chemical inhibitors and siRNA targeted knockdown, we found that NOX1, NOX2, NOX4, and XO-derived ROS regulates induction of proinflammatory mediator levels. Like airway epithelial cells in vitro, NOX inhibitor VAS2870 reduced keratinocyte chemoattractant (KC), IL-6, and TNF-α production and 4-hydroxynonenal (4-HNE) staining induced by dust extract in mouse lungs. VAS2870 inhibition of proinflammatory mediators was associated with reduced NFκB and Stat3 activation indicating that NOX generated ROS activates NFκB and Stat3 to induce proinflammatory gene expression. Dust extract increased the membrane association of p47phox in airway epithelial cells indicating NOX2 activation but had no effect on NOX2 protein levels. In summary, our studies have shown that NOX and XO generated ROS control organic dust induction of proinflammatory mediators in airway epithelial cells via NFκB and Stat3 activation.
Sigma-Aldrich
Anti-p47 PHOX antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.