Merck
IgA Antibodies and IgA Deficiency in SARS-CoV-2 Infection.
Frontiers in cellular and infection microbiology (2021-04-24)
Isabella Quinti, Eva Piano Mortari, Ane Fernandez Salinas, Cinzia Milito, Rita Carsetti
ABSTRACT
A large repertoire of IgA is produced by B lymphocytes with T-independent and T-dependent mechanisms useful in defense against pathogenic microorganisms and to reduce immune activation. IgA is active against several pathogens, including rotavirus, poliovirus, influenza virus, and SARS-CoV-2. It protects the epithelial barriers from pathogens and modulates excessive immune responses in inflammatory diseases. An early SARS-CoV-2 specific humoral response is dominated by IgA antibodies responses greatly contributing to virus neutralization. The lack of anti-SARS-Cov-2 IgA and secretory IgA (sIgA) might represent a possible cause of COVID-19 severity, vaccine failure, and possible cause of prolonged viral shedding in patients with Primary Antibody Deficiencies, including patients with Selective IgA Deficiency. Differently from other primary antibody deficiency entities, Selective IgA Deficiency occurs in the vast majority of patients as an asymptomatic condition, and it is often an unrecognized, Studies are needed to clarify the open questions raised by possible consequences of a lack of an IgA response to SARS-CoV-2.
Sigma-Aldrich
IgA from human serum, reagent grade, ≥95% (HPLC), buffered aqueous solution