Merck
  • Universal non-antibody detection of protein phosphorylation using pIMAGO.
Universal non-antibody detection of protein phosphorylation using pIMAGO.
Current protocols in chemical biology (2015-03-03)
Anton B Iliuk, W Andy Tao
ABSTRACT
This article describes methods for a new, non-antibody phosphorylation detection reagent, termed pIMAGO (phospho-imaging). This novel reagent takes advantage not only of the unique properties of the soluble nanoparticles, but also of the multiple functionalities of the molecule, allowing for highly selective, sensitive, and quantitative assessment of protein phosphorylation without using radioactive isotopes or phospho-specific antibodies. The methods allow for multiplexed detection of phosphorylation and total protein amount simultaneously. The straightforward and routine detection and quantitation of general phosphorylation on any site of any protein can be performed in western blot and ELISA formats.
Sigma-Aldrich
Avidin–Peroxidase, lyophilized powder