Merck
L-2-aminobutyric acid: two fully ordered polymorphs with Z' = 4.
Acta crystallographica. Section B, Structural science (2010-03-23)
Carl Henrik Görbitz
ABSTRACT
The crystal structure of L-2-aminobutyric acid, an L-alanine analogue with an ethyl rather than a methyl side chain, has proved elusive owing to problems growing diffraction quality crystals. Good diffraction data have now been obtained for two polymorphs, in space groups P2(1) and I2, revealing surprisingly complex, yet fully ordered crystalline arrangements with Z' = 4. The closely related structures are divided into hydrophilic and hydrophobic layers, the latter being the thinnest ever found for an amino acid (other than alpha-glycine). The hydrophobic layers furthermore contain conspicuous pseudo-centers-of-symmetry, leading to overall centrosymmetric intensity statistics. Uniquely, the four molecules in the asymmetric unit can be divided into two pairs that each forms an independent hydrogen-bond network.
Sigma-Aldrich
L-2-Aminobutyric acid, ≥99% (titration)
Pricing and availability is not currently available.