Merck
  • Serotonin uptake inhibitors: uses in clinical therapy and in laboratory research.
Serotonin uptake inhibitors: uses in clinical therapy and in laboratory research.
Progress in drug research. Fortschritte der Arzneimittelforschung. Progres des recherches pharmaceutiques (1995-01-01)
R W Fuller
ABSTRACT
Fluoxetine, zimelidine, sertraline, paroxetine, fluvoxamine, indalpine and citalopram are the selective inhibitors of serotonin uptake that have been most widely studied. Some of these compounds are or have been used clinically in the treatment of mental depression, obsessive-compulsive disorder and bulimia, and therapeutic benefit has been claimed in additional diseases as well. By blocking the membrane uptake carrier which transports serotonin from the extracellular space to inside the serotonin nerve terminals, these compounds increase extracellular concentrations of serotonin and amplify signals sent by serotonin neurons. Because serotonin neurons are widespread in the central nervous system, the functional consequences of blocking serotonin uptake are diverse, but are generally subtle. Animals treated with serotonin uptake inhibitors look normal in gross appearance, but effects such as reduced aggressive behavior, decreased food intake and altered food selection, analgesia, anticonvulsant activity, endocrine changes and neurochemical changes have been demonstrated and characterized. Serotonin uptake inhibitors have helped in revealing some dynamics of serotonin neurons; for example, when uptake is inhibited and extracellular serotonin concentration increases, presynaptic as well as postsynaptic receptors for serotonin are activated to a greater degree. A consequence of increased activation of autoreceptors on serotonin cell bodies and nerve terminals is a reduction in firing of serotonin neurons and a decrease in serotonin synthesis and release. The result is a limit on the degree to which extracellular serotonin and serotonergic neurotransmission are increased.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Zimelidine dihydrochloride, solid
Pricing and availability is not currently available.