Merck
  • A multi-dimensional, time-lapse, high content screening platform applied to schistosomiasis drug discovery.
A multi-dimensional, time-lapse, high content screening platform applied to schistosomiasis drug discovery.
Communications biology (2020-12-23)
Steven Chen, Brian M Suzuki, Jakob Dohrmann, Rahul Singh, Michelle R Arkin, Conor R Caffrey
ABSTRACT
Approximately 10% of the world's population is at risk of schistosomiasis, a disease of poverty caused by the Schistosoma parasite. To facilitate drug discovery for this complex flatworm, we developed an automated high-content screen to quantify the multidimensional responses of Schistosoma mansoni post-infective larvae (somules) to chemical insult. We describe an integrated platform to process worms at scale, collect time-lapsed, bright-field images, segment highly variable and touching worms, and then store, visualize, and query dynamic phenotypes. To demonstrate the methodology, we treated somules with seven drugs that generated diverse responses and evaluated 45 static and kinetic response descriptors relative to concentration and time. For compound screening, we used the Mahalanobis distance to compare multidimensional phenotypic effects induced by 1323 approved drugs. Overall, we characterize both known anti-schistosomals and identify new bioactives. Apart from facilitating drug discovery, the multidimensional quantification provided by this platform will allow mapping of chemistry to phenotype.
Sigma-Aldrich
Imipramine hydrochloride, BioXtra, ≥99% (TLC)
Pricing and availability is not currently available.