Merck
  • Temperature-Responsive Pyraclostrobin-Loaded Octadecane Submicrocapsules with Lowered Toxicity.
Temperature-Responsive Pyraclostrobin-Loaded Octadecane Submicrocapsules with Lowered Toxicity.
Nanomaterials (Basel, Switzerland) (2020-12-03)
Aiym Tleuova, Vssl Prasad Talluri, Rohitesh Kumar, František Štěpánek
ABSTRACT
Pyraclostrobin (Pyr) is one of the most effective fungicides. However, it can degrade via photolysis in water, it is toxic to aquatic life and if inhaled, it has a low solubility in water, that leads to difficulties when applying to plants by spraying. Additionally, the necessity of repeated (weekly) sprays of fungicides when the pathogen growth risk is the highest, such as at the temperature range of 24 to 36 °C and increased humidity of about 95%, leads to loss of efficiency of the fungicide and overdose of chemicals. In the present study, pyraclostrobin was microencapsulated to solve the abovementioned issues. As a core of capsules octadecane (OD) with a melting point of 28 °C was used, thus, the release of pyraclostrobin was controlled via temperature change. Pyraclostrobin-loaded submicrocapsules (PyrSMCs) were characterized using SEM, DLS, TGA/DSC, HPLC, FTIR methods; stimuli-responsivity was tested employing in vitro tests with pathogenic culture (Fungal strain of Pyrenophora teres - CPPF-453) grown in Petri dishes. Toxicity of PyrSMCs to Artemia salina was studied as well. Size of capsules was 200-600 nm along with the presence of bigger capsules with a diameter of 1-4 µm. PyrSMCs showed excellent antifungal effects above the melting point of octadecane. PyrSMCs demonstrated 29 times less toxicity than pyraclostrobin of technical grade. Overall, results show the potential of such capsules to be applied in the agricultural industry for precise agriculture strategies.
Sigma-Aldrich
Octadecane, 99%
Pricing and availability is not currently available.