Merck
  • Nitrocellulose-bound achromopeptidase for point-of-care nucleic acid tests.
Nitrocellulose-bound achromopeptidase for point-of-care nucleic acid tests.
Scientific reports (2021-03-19)
Georgios Chondrogiannis, Shirin Khaliliazar, Anna Toldrà, Pedro Réu, Mahiar M Hamedi
ABSTRACT
Enzymes are the cornerstone of modern biotechnology. Achromopeptidase (ACP) is a well-known enzyme that hydrolyzes a number of proteins, notably proteins on the surface of Gram-positive bacteria. It is therefore used for sample preparation in nucleic acid tests. However, ACP inhibits DNA amplification which makes its integration difficult. Heat is commonly used to inactivate ACP, but it can be challenging to integrate heating into point-of-care devices. Here, we use recombinase polymerase amplification (RPA) together with ACP, and show that when ACP is immobilized on nitrocellulose paper, it retains its enzymatic function and can easily and rapidly be activated using agitation. The nitrocellulose-bound ACP does, however, not leak into the solution, preventing the need for deactivation through heat or by other means. Nitrocellulose-bound ACP thus opens new possibilities for paper-based Point-of-Care (POC) devices.
Sigma-Aldrich
Achromopeptidase from bacteria, lyophilized powder, ≥1,000 units/mg solid
Pricing and availability is not currently available.