Merck
  • Heterogeneous disease-propagating stem cells in juvenile myelomonocytic leukemia.
Heterogeneous disease-propagating stem cells in juvenile myelomonocytic leukemia.
The Journal of experimental medicine (2021-01-09)
Eleni Louka, Benjamin Povinelli, Alba Rodriguez-Meira, Gemma Buck, Wei Xiong Wen, Guanlin Wang, Nikolaos Sousos, Neil Ashley, Angela Hamblin, Christopher A G Booth, Anindita Roy, Natalina Elliott, Deena Iskander, Josu de la Fuente, Nicholas Fordham, Sorcha O'Byrne, Sarah Inglott, Ruggiero Norfo, Mariolina Salio, Supat Thongjuea, Anupama Rao, Irene Roberts, Adam J Mead
ABSTRACT
Juvenile myelomonocytic leukemia (JMML) is a poor-prognosis childhood leukemia usually caused by RAS-pathway mutations. The cellular hierarchy in JMML is poorly characterized, including the identity of leukemia stem cells (LSCs). FACS and single-cell RNA sequencing reveal marked heterogeneity of JMML hematopoietic stem/progenitor cells (HSPCs), including an aberrant Lin-CD34+CD38-CD90+CD45RA+ population. Single-cell HSPC index-sorting and clonogenic assays show that (1) all somatic mutations can be backtracked to the phenotypic HSC compartment, with RAS-pathway mutations as a "first hit," (2) mutations are acquired with both linear and branching patterns of clonal evolution, and (3) mutant HSPCs are present after allogeneic HSC transplant before molecular/clinical evidence of relapse. Stem cell assays reveal interpatient heterogeneity of JMML LSCs, which are present in, but not confined to, the phenotypic HSC compartment. RNA sequencing of JMML LSC reveals up-regulation of stem cell and fetal genes (HLF, MEIS1, CNN3, VNN2, and HMGA2) and candidate therapeutic targets/biomarkers (MTOR, SLC2A1, and CD96), paving the way for LSC-directed disease monitoring and therapy in this disease.
Sigma-Aldrich
Fasentin, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade