Merck
  • LOXL1 confers antiapoptosis and promotes gliomagenesis through stabilizing BAG2.
LOXL1 confers antiapoptosis and promotes gliomagenesis through stabilizing BAG2.
Cell death and differentiation (2020-05-20)
Hua Yu, Jun Ding, Hongwen Zhu, Yao Jing, Hu Zhou, Hengli Tian, Ke Tang, Gang Wang, Xiongjun Wang
ABSTRACT
The lysyl oxidase (LOX) family is closely related to the progression of glioma. To ensure the clinical significance of LOX family in glioma, The Cancer Genome Atlas (TCGA) database was mined and the analysis indicated that higher LOXL1 expression was correlated with more malignant glioma progression. The functions of LOXL1 in promoting glioma cell survival and inhibiting apoptosis were studied by gain- and loss-of-function experiments in cells and animals. LOXL1 was found to exhibit antiapoptotic activity by interacting with multiple antiapoptosis modulators, especially BAG family molecular chaperone regulator 2 (BAG2). LOXL1-D515 interacted with BAG2-K186 through a hydrogen bond, and its lysyl oxidase activity prevented BAG2 degradation by competing with K186 ubiquitylation. Then, we discovered that LOXL1 expression was specifically upregulated through the VEGFR-Src-CEBPA axis. Clinically, the patients with higher LOXL1 levels in their blood had much more abundant BAG2 protein levels in glioma tissues. Conclusively, LOXL1 functions as an important mediator that increases the antiapoptotic capacity of tumor cells, and approaches targeting LOXL1 represent a potential strategy for treating glioma. In addition, blood LOXL1 levels can be used as a biomarker to monitor glioma progression.