Merck
Androgen-dependent regulation of Her-2/neu in prostate cancer cells.
Cancer research (2006-06-03)
Raanan Berger, Douglas I Lin, Maria Nieto, Ewa Sicinska, Levi A Garraway, Heiner Adams, Sabina Signoretti, William C Hahn, Massimo Loda
ABSTRACT
The mechanisms underlying the progression of prostate cancer to a state of resistance to hormone ablation remain poorly understood. Here, we have investigated the relationship between androgen receptor (AR) and Her-2/neu in prostate cancer cells. Overexpression of Her-2/neu (c-ErbB2) activates the AR pathway and confers a survival and growth advantage to prostate cancer cells in an androgen-deficient milieu. In vitro, the absence of androgens or AR blockade induced Her-2/neu protein expression and phosphorylation. In contrast, upon readministration of androgens, Her-2/neu mRNA, protein, and phosphorylation levels decreased linearly with increasing concentrations of dihydrotestosterone as LNCaP cells reentered the cell cycle. In vivo, induction of Her-2/neu by castration in orthotopically injected LNCaP cells resulted in a progressive increase in prostate-specific antigen secretion into the mouse serum, indicating that Her-2/neu-mediated, AR-dependent transcription occurs following castration and results in tumor cell growth. Finally, selection of LNCaP cells stably transfected with short hairpin RNA specific for AR resulted in Her-2/neu overexpression. Similarly, knockdown of Her-2/neu led to induction of AR. However, when Her-2/neu and AR were simultaneously targeted, we observed cell death, whereas surviving cells retained low level expression of Her-2/neu. Thus, induction and activation of Her-2/neu occurs in an androgen-depleted environment or as a result of AR inactivation, promoting ablation-resistant survival of prostate cancer cells. These data provide the biochemical rationale to target Her-2/neu in hormone-refractory prostate cancer.
Sigma-Aldrich
Anti-Cyclin D1 (Ab-3) Mouse mAb (DCS-6), liquid, clone DCS-6, Calbiochem®