Merck
Expression of Notch1 and Jagged2 in the enteric nervous system.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2003-06-18)
Guy R Sander, Simon J H Brookes, Barry C Powell
ABSTRACT
The Notch signaling pathway is a vitally important pathway in regulating brain development. To explore the involvement of the Notch pathway in neuronal cells of adult rat gut, we investigated the expression of Notch1 and Jagged2 by in situ hybridization (ISH) and immunohistochemistry (IHC). In the enteric nervous system, Notch1 and Jagged2 were expressed in ganglia of the submucosal and myenteric plexus. Notch1 was preferentially expressed in cholinergic neurons lacking calretinin or nitric oxide synthase (NOS), whereas Jagged2 was present in most neuron subtypes. We propose that Notch1 and Jagged2 have a continuing role in the maintenance and function of neuronal cells in the adult enteric nervous system.
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, serum, Chemicon®