Merck
  • Dietary Supplementation of Probiotic Lactobacillus acidophilus Modulates Cholesterol Levels, Immune Response, and Productive Performance of Laying Hens.
Dietary Supplementation of Probiotic Lactobacillus acidophilus Modulates Cholesterol Levels, Immune Response, and Productive Performance of Laying Hens.
Animals : an open access journal from MDPI (2020-09-10)
Abdulaziz A Alaqil, Ahmed O Abbas, Hossam S El-Beltagi, Hanaa K Abd El-Atty, Gamal M K Mehaisen, Eman S Moustafa
ABSTRACT
This study examines the effect of dietary supplementation with Lactobacillus acidophilus (LA) on the cholesterol levels, immune response, and productive performance of laying hens. A total of 216, 40-week-old, commercial Hy-Line brown chicken layers were randomly assigned into four treatment groups (18 birds × three replicates per group) and fed diet supplemented with 0 (control), 1 × 109, 21 × 109, and 31 × 109 colony forming units (CFUs) of Lactobacillus acidophilus (LA) per kg of feed for six consecutive weeks. Results show that plasma triglycerides, low-density lipoprotein (LDL) and total cholesterols became lesser, while high-density lipoprotein (HDL) cholesterol became higher in LA-supplemented groups compared to the control. In addition, a significant reduction occurred in the liver and egg yolk cholesterol by LA supplementation. Moreover, the immunological parameters including antibody titer against sheep red blood cells (SRBCs), phytohemagglutinin (PHA)-wattle swelling test, and T- & B-lymphocyte proliferation were enhanced in laying hens supplemented with LA compared to the control hens. While the heterophil to lymphocyte (H/L) ratio decreased with LA supplementation, indicating low stress conditions in the treated hens. These positive effects for LA were further reflected on the productive performance of laying hens and improved egg production, egg weight, egg mass, and feed efficiency. Our findings indicate that LA probiotic could be recommended in laying hens' diets for lowering egg yolk cholesterol with positive impacts on health and performance.
Sigma-Aldrich
5-α-Cholestane, ≥97.0% (HPLC)
Pricing and availability is not currently available.