Merck
  • The O-fucose glycan in the ligand-binding domain of Notch1 regulates embryogenesis and T cell development.
The O-fucose glycan in the ligand-binding domain of Notch1 regulates embryogenesis and T cell development.
Proceedings of the National Academy of Sciences of the United States of America (2008-01-30)
Changhui Ge, Pamela Stanley
ABSTRACT
Mechanisms by which the extracellular domain of Notch1 controls Notch1 signaling are not well defined. Here, we show that the O-fucose glycan in the Notch1 ligand-binding domain regulates the strength of Notch1 signaling during embryogenesis, postweaning growth, and T cell development in the mouse. Heterozygotes carrying a Notch1(12f) allele and an inactive Notch1 allele die at approximately embryonic day (E)12 with a typical Notch1 null phenotype. Homozygous Notch1(12f/12f) mice are viable and fertile but grow somewhat more slowly than littermates after weaning. Notch1(12f/12f) thymocytes bind less Delta1 and exhibit reduced Notch1 signaling. The number of double-positive (DP) and single-positive (SP) T cells are decreased in Notch1(12f/12f) thymus, and DP T cells are more apoptotic. By contrast, proportionately more SP cells have matured, and SP-to-DP ratios are increased in mutant thymus. Thus, the O-fucose glycan in EGF12 of mouse Notch1 is required for optimal Notch1 signaling and T cell development in mammals.
Sigma-Aldrich
Anti-Notch 1 Antibody, extracellular, clone 8G10, culture supernatant, clone 8G10, Chemicon®