Merck
  • Negative Regulation of Tec Kinase Alleviates LPS-Induced Acute Kidney Injury in Mice via theTLR4/NF-κB Signaling Pathway.
Negative Regulation of Tec Kinase Alleviates LPS-Induced Acute Kidney Injury in Mice via theTLR4/NF-κB Signaling Pathway.
BioMed research international (2020-07-21)
Wei Zhang, Ping Zhou, Xiao Jiang, Zhe Fan, Xingxin Xu, Fei Wang
ABSTRACT
Tec kinase is an important mediator in inflammatory immune response that enhances the activity of neutrophils and macrophages. However, information on its function in lipopolysaccharide- (LPS-) induced acute kidney injury (AKI) is limited. This study is aimed at determining whether Tec kinase was a regulator in AKI. An AKI model in mice was successfully established using intraperitoneal LPS. Results showed that the serum levels of creatinine (Cr), blood urea nitrogen (BUN), and cystatin-C (Cys-C) increased after intraperitoneal LPS injection. Renal tissue sustained significantly severe injury as measured by pathological scores. Pretreatment with LFM-A13 improved the function of the kidney in mice and decreased the renal injury score. Enzyme-linked immunosorbent assay showed that LFM-A13 significantly reduced the release of IL-1β and TNF-α in mice exposed to LPS. LFM-A13 can evidently abrogate the expression of Tec protein, MyD88, TLR4, NF-κB p65, and Tec's phosphorylated protein as determined by Western blot. Immunohistochemistry analysis revealed that LFM-A13 markedly downregulated the expression of Tec kinase in renal tubular epithelial cells. In vitro, Tec kinase protein was expressed highly in NRK-52E cells after LPS exposure. Tec-siRNA also decreased IL-1β and TNF-α production and obviously abolished phospho-p65 and phospho-IκBα expression in NRK-52E cell stimulated by LPS; however, Tec-siRNA increased the IκBα level. Altogether, these data suggested that Tec kinase can be a modulating protein in AKI through TLR4/NF-κB activation.
Sigma-Aldrich
LFM-A13, ≥98% (HPLC), powder
Pricing and availability is not currently available.