Merck
  • TRIB3-EGFR interaction promotes lung cancer progression and defines a therapeutic target.
TRIB3-EGFR interaction promotes lung cancer progression and defines a therapeutic target.
Nature communications (2020-07-23)
Jiao-Jiao Yu, Dan-Dan Zhou, Xiao-Xiao Yang, Bing Cui, Feng-Wei Tan, Junjian Wang, Ke Li, Shuang Shang, Cheng Zhang, Xiao-Xi Lv, Xiao-Wei Zhang, Shan-Shan Liu, Jin-Mei Yu, Feng Wang, Bo Huang, Fang Hua, Zhuo-Wei Hu
ABSTRACT
High expression or aberrant activation of epidermal growth factor receptor (EGFR) is related to tumor progression and therapy resistance across cancer types, including non-small cell lung cancer (NSCLC). EGFR tyrosine kinase inhibitors (TKIs) are first-line therapy for NSCLC. However, patients eventually deteriorate after inevitable acquisition of EGFR TKI-resistant mutations, highlighting the need for therapeutics with alternative mechanisms of action. Here, we report that the elevated tribbles pseudokinase 3 (TRIB3) is positively associated with EGFR stability and NSCLC progression. TRIB3 interacts with EGFR and recruits PKCα to induce a Thr654 phosphorylation and WWP1-induced Lys689 ubiquitination in the EGFR juxtamembrane region, which enhances EGFR recycling, stability, downstream activity, and NSCLC stemness. Disturbing the TRIB3-EGFR interaction with a stapled peptide attenuates NSCLC progression by accelerating EGFR degradation and sensitizes NSCLC cells to chemotherapeutic agents. These findings indicate that targeting EGFR degradation is a previously unappreciated therapeutic option in EGFR-related NSCLC.
Sigma-Aldrich
Human EGFR / ErbB1 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Pricing and availability is not currently available.