Merck
  • Identification of a fructose-1,6-bisphosphate aldolase gene and association of the single nucleotide polymorphisms with growth traits in the clam Meretrix meretrix.
Identification of a fructose-1,6-bisphosphate aldolase gene and association of the single nucleotide polymorphisms with growth traits in the clam Meretrix meretrix.
Molecular biology reports (2011-12-14)
Chao Wang, Hongxia Wang, Yan Li, Baozhong Liu
ABSTRACT
This study investigated whether there were single nucleotide polymorphisms (SNPs) in fructose-1,6-bisphosphate aldolase (FBA) gene associated with growth traits of the clam Meretrix meretrix. A FBA gene was identified in M. meretrix and its deduced amino acid residues shared high identity with type I aldolase. The FBA (MmeFBA) mRNA expression profile was examined by real-time PCR in different tissues and the significantly high expression level in foot and adduct muscle suggests that MmeFBA is a muscle type aldolase which functions in glycolytic pathway. In the MmeFBA gene, we identified four intron SNPs and three exon SNPs including a nonsynonymous SNP (mmfbae-2). These SNPs were genotyped in 205 clams from two clam populations with significantly different growth performance. Results showed that allele frequencies of three SNPs (mmfbai-1, mmfbai-3 and mmfbae-2) and the genotype frequency of mmfbai-1 were all significantly different between the two populations. The haplotype analysis further supported the three SNPs distributed differently between the two populations. This study successively characterized three growth-related SNPs in a gene involved in energy metabolism of M. meretrix. These findings could contribute the development of phenotype-selective breeding program in M. meretrix.
Sigma-Aldrich
D-Fructose 1,6-bisphosphate tetra(cyclohexylammonium) salt, ≥95% anhydrous basis (enzymatic)
Pricing and availability is not currently available.