Merck
  • Formulation and evaluation of mucoadhesive buccal films of enalapril maleate.
Formulation and evaluation of mucoadhesive buccal films of enalapril maleate.
Indian journal of pharmaceutical sciences (2011-06-23)
A Semalty, Mona Semalty, U Nautiyal
ABSTRACT
Enalapril maleate is used in the treatment of hypertension and angina pectoris. It shows low bioavailability due to high hepatic first pass metabolism. Hence the present work was undertaken to formulate mucoadhesive buccal films of enalapril maleate with an objective to improve therapeutic efficacy, patient compliance and the bioavailability. In the present study ten formulations of mucoadhesive drug delivery system of enalapril maleate were prepared as buccal films, by solvent casting technique. Sodium carboxymethylcellulose, hydroxylpropylmethylcellulose, hydroxyethylcellulose and polyvinyl pyrrolidone K-90 were used as mucoadhesive polymers. Prepared films were evaluated for their weight, thickness, surface pH, swelling index, drug content uniformity, in vitro residence time, folding endurance in vitro release and permeation studies. Films exhibited controlled release over more than 10 h in permeation studies. It was concluded that the films containing 20 mg of enalapril maleate in sodium carboxymethylcellulose 2% w/v and hydroxyethyl cellulose 2% w/v (formulation F5), showed good swelling, a convenient residence time and promising controlled drug release, thus can be selected for the development of buccal film for effective therapeutic uses.
Sigma-Aldrich
Hydroxyethyl-cellulose, viscosity 90-160 cP, 5 % in H2O(25 °C)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxyethyl-cellulose
Pricing and availability is not currently available.