Merck
  • Plasma exosomal prion protein levels are correlated with cognitive decline in PD patients.
Plasma exosomal prion protein levels are correlated with cognitive decline in PD patients.
Neuroscience letters (2020-02-29)
Bing Leng, Hairong Sun, Junwu Zhao, Yuanyuan Liu, Tengqun Shen, Wenjin Liu, Xiaoxiao Liu, Ming Tan, Fang Li, Jinbiao Zhang, Zhenguang Li
ABSTRACT
Cognitive decline is a common non-motor symptom of Parkinson disease (PD), and cellular prion protein (PrPC) has been suggested to play a role in this process. This study aimed to investigate the correlation between plasma exosomal prion protein and cognitive decline in PD patients. A total of 60 participants, which included 23 PD patients without cognitive impairment (the PD-NCI group), 17 PD patients with cognitive impairment (the PD-CI group) and 20 health controls were included in this study. All participants received a complete evaluation of motor symptoms as well as non-motor symptoms, which include devaluations of cognitive function(assessed with the Montreal Cognitive Assessment (MoCA)) and their psychiatric state(assessed with the Hamilton Anxiety Scale(HAM-A) and Hamilton Depression Scale(HAMD-17)). We used an enzyme-linked immunosorbent assay (ELISA) to measure the plasma exosomal prion protein level. The exosomal marker Heat shock protein 70 (HSP 70) was used to normalize the protein level to the exosome content. In PD patients, the plasma exosomal prion protein concentration was negatively correlated with the cognitive level. The plasma exosomal prion protein concentration was significantly higher in the PD-CI group than in the control group (p < 0.05) and the PD-NCI group (p < 0.05).Multivariate regression analysis indicated that plasma exosomal prion protein levels were significantly associated with the cognitive level (t=-3.185, P = 0.001) after adjusting for age, education, disease duration, H&Y stage and MDS-UPDRS-III scores. The plasma exosomal prion protein level is correlated with cognitive decline in PD patients and might be a potential biomarker for PD patients at risk for cognitive impairment.
Sigma-Aldrich
Human HSP70 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Pricing and availability is not currently available.