Merck
  • Preparative separation of liquiritigenin and glycyrrhetic acid from Glycyrrhiza uralensis Fisch using hydrolytic extraction combined with high-speed countercurrent chromatography.
Preparative separation of liquiritigenin and glycyrrhetic acid from Glycyrrhiza uralensis Fisch using hydrolytic extraction combined with high-speed countercurrent chromatography.
Biomedical chromatography : BMC (2020-01-04)
Hao Wang, Hu Shan, Haitao Lü
ABSTRACT
The objective of this paper was to develop a preparative method for the isolation and purification of liquiritigenin and glycyrrhetic acid from Glycyrrhiza uralensis Fisch using hydrolytic extraction combined with high-speed countercurrent chromatography (HSCCC). Liquiritigenin and glycyrrhetic acid were well hydrolyzed from liquiritin and glycyrrhizic acid by hydrochloric acid, respectively. The optimal extraction conditions were obtained by single-factor and orthogonal experiments, which were 100% ethanol, 1.5 mol/L hydrochloric acid, 1:25 ratio of solid to liquid, and extracted 2 h for one time. Using the two-phase solvent system of n-hexane-ethyl acetate-methanol-water (4:5:4:5, v/v), 2.1 mg liquiritigenin (the purity was 96.5% with a recovery of 87.6%) and 12.3 mg glycyrrhetic acid (the purity was 97.1% with a recovery of 74.4%) were obtained from 315-mg crude extraction by HSCCC. The retention ratio of stationary phase was 47.2%. Their structures were identified by HPLC, melting points, UV, Fourier-transform infrared, Electrospray ionization-MS, 1 H nuclear magnetic resonance (NMR), and 13 C NMR spectra. According to the antioxidant activity assays, liquiritigenin and glycyrrhetic acid had some scavenging abilities on 1,1-diphenyl-2-picrylhydrazyl free radicals; liquiritigenin had stronger scavenging ability on hydroxyl radicals.
Product Description
Liquiritigenin, phyproof® Reference Substance
Sigma-Aldrich
Liquiritigenin, ≥97.0% (HPLC)