Merck
  • Double diastereoselection in aldol reactions mediated by dicyclohexylchloroborane between L-erythrulose derivatives and chiral aldehydes. The Felkin-Anh versus Cornforth dichotomy.
Double diastereoselection in aldol reactions mediated by dicyclohexylchloroborane between L-erythrulose derivatives and chiral aldehydes. The Felkin-Anh versus Cornforth dichotomy.
The Journal of organic chemistry (2003-10-25)
J Alberto Marco, Miguel Carda, Santiago Díaz-Oltra, Juan Murga, Eva Falomir, Harald Roeper
ABSTRACT
Both matched and mismatched diastereoselections have been observed in aldol reactions of the B,B-dicyclohexylboron enolate of a protected l-erythrulose derivative with a range of chiral aldehydes. The stereochemical outcome of reactions with alpha-methyl aldehydes can be adequately explained within the Felkin-Anh paradigm. In the case of alpha-oxygenated aldehydes, however, strict adherence to this model does not allow for a satisfactory account of the observed results. In such cases, the Cornforth model provides a much better explanation.
Sigma-Aldrich
L-(+)-Erythrulose, ≥75% (HPLC)
Pricing and availability is not currently available.