Merck
  • IGF2BP3 functions as a potential oncogene and is a crucial target of miR-34a in gastric carcinogenesis.
IGF2BP3 functions as a potential oncogene and is a crucial target of miR-34a in gastric carcinogenesis.
Molecular cancer (2017-04-13)
Yuhang Zhou, Tingting Huang, Ho Lam Siu, Chi Chun Wong, Yujuan Dong, Feng Wu, Bin Zhang, William K K Wu, Alfred S L Cheng, Jun Yu, Ka Fai To, Wei Kang
ABSTRACT
Gastric cancer (GC) is one of the frequent causes of cancer-related death in eastern Asian population. IGF2BP2 lists in the top rank up-regulated genes in GC, but its functional role is unclear. The expression of IGF2BP3 in GC cell lines and primary samples was examined by qRT-PCR and Western blot. The biological role of IGF2BP3 was revealed by a series of functional in vitro studies. Its regulation by microRNAs (miRNAs) was predicted by TargetScan and confirmed by luciferase assays and rescue experiments. IGF2BP3 ranked the No.1 of the up-regulated genes by expression microarray analysis in GC cell lines. The expression level of IGF2BP3 was observed in GC tissues comparing with non-tumorous gastric epitheliums. The up-regulated IGF2BP3 expression was associated with poor disease specific survival. IGF2BP3 knockdown significantly inhibited cell proliferation and invasion. Apart from copy number gain, IGF2BP3 has been confirmed to be negatively regulated by tumor-suppressive miRNA, namely miR-34a. The expression of miR-34a showed negative correlation with IGF2BP3 mRNA expression in primary GC samples and more importantly, re-overexpression of IGF2BP3 rescued the inhibitory effect of miR-34a. We compressively revealed the oncogenic role of IGF2BP3 in gastric tumorigenesis and confirmed its activation is partly due to the silence of miR-34a. Our findings identified useful prognostic biomarker and provided clinical translational potential.
Sigma-Aldrich
Anti-IGF2 mRNA-binding protein 3 Antibody, from rabbit, purified by affinity chromatography