Merck
  • A biomaterial with a channel-like pore architecture induces endochondral healing of bone defects.
A biomaterial with a channel-like pore architecture induces endochondral healing of bone defects.
Nature communications (2018-10-27)
A Petersen, A Princ, G Korus, A Ellinghaus, H Leemhuis, A Herrera, A Klaumünzer, S Schreivogel, A Woloszyk, K Schmidt-Bleek, S Geissler, I Heschel, G N Duda
ABSTRACT
Biomaterials developed to treat bone defects have classically focused on bone healing via direct, intramembranous ossification. In contrast, most bones in our body develop from a cartilage template via a second pathway called endochondral ossification. The unsolved clinical challenge to regenerate large bone defects has brought endochondral ossification into discussion as an alternative approach for bone healing. However, a biomaterial strategy for the regeneration of large bone defects via endochondral ossification is missing. Here we report on a biomaterial with a channel-like pore architecture to control cell recruitment and tissue patterning in the early phase of healing. In consequence of extracellular matrix alignment, CD146+ progenitor cell accumulation and restrained vascularization, a highly organized endochondral ossification process is induced in rats. Our findings demonstrate that a pure biomaterial approach has the potential to recapitulate a developmental bone growth process for bone healing. This might motivate future strategies for biomaterial-based tissue regeneration.
Sigma-Aldrich
Anti-Nerve Growth Factor Receptor Antibody, extracellular, clone 192-IgG, clone 192-IgG, Chemicon®, from mouse