Merck
  • Terminalia chebula (fruit) prevents liver toxicity caused by sub-chronic administration of rifampicin, isoniazid and pyrazinamide in combination.
Terminalia chebula (fruit) prevents liver toxicity caused by sub-chronic administration of rifampicin, isoniazid and pyrazinamide in combination.
Human & experimental toxicology (2006-04-26)
S A Tasduq, K Singh, N K Satti, D K Gupta, K A Suri, R K Johri
ABSTRACT
Terminalia chebula Gertn. (Combetraceae) is an important herbal drug in Ayurvedic pharmacopea. In the present study, a 95% ethanolic extract of T. chebula (fruit) (TC extract), which was chemically characterized on the basis of chebuloside II as a marker, was investigated for hepatoprotective activity against anti-tuberculosis (anti-TB) drug-induced toxicity. TC extract was found to prevent the hepatotoxicity caused by the administration of rifampicin (RIF), isoniazid (INH) and pyrazinamide (PZA) (in combination) in a sub-chronic mode (12 weeks). The hepatoprotective effect of TC extract could be attributed to its prominent anti-oxidative and membrane stabilizing activities. The changes in biochemical observations were supported by histological profile.
Sigma-Aldrich
Pyrazinecarboxamide