Merck
  • Ruthenium half-sandwich complexes as protein kinase inhibitors: an N-succinimidyl ester for rapid derivatizations of the cyclopentadienyl moiety.
Ruthenium half-sandwich complexes as protein kinase inhibitors: an N-succinimidyl ester for rapid derivatizations of the cyclopentadienyl moiety.
Organic letters (2006-11-17)
Howard Bregman, Eric Meggers
ABSTRACT
Cyclopentadienyl half-sandwich ruthenium complexes have been demonstrated to be promising scaffolds as protein kinase inhibitors. In order to rapidly identify derivatives which display modified pharmacological properties, we developed the synthesis of an organoruthenium compound bearing an N-succinimidyl ester at the cyclopentadienyl moiety. The quenching of this activated ester with a library of primary amines, followed by testing of the resulting amide library, led to the identification of organometallic Pim-1 and GSK-3 inhibitors with improved potencies and kinase selectivities. [structure: see text].