Merck
  • Immunization expands B cells specific to HIV-1 V3 glycan in mice and macaques.
Immunization expands B cells specific to HIV-1 V3 glycan in mice and macaques.
Nature (2019-05-31)
Amelia Escolano, Harry B Gristick, Morgan E Abernathy, Julia Merkenschlager, Rajeev Gautam, Thiago Y Oliveira, Joy Pai, Anthony P West, Christopher O Barnes, Alexander A Cohen, Haoqing Wang, Jovana Golijanin, Daniel Yost, Jennifer R Keeffe, Zijun Wang, Peng Zhao, Kai-Hui Yao, Jens Bauer, Lilian Nogueira, Han Gao, Alisa V Voll, David C Montefiori, Michael S Seaman, Anna Gazumyan, Murillo Silva, Andrew T McGuire, Leonidas Stamatatos, Darrell J Irvine, Lance Wells, Malcolm A Martin, Pamela J Bjorkman, Michel C Nussenzweig
ABSTRACT
Broadly neutralizing monoclonal antibodies protect against infection with HIV-1 in animal models, suggesting that a vaccine that elicits these antibodies would be protective in humans. However, it has not yet been possible to induce adequate serological responses by vaccination. Here, to activate B cells that express precursors of broadly neutralizing antibodies within polyclonal repertoires, we developed an immunogen, RC1, that facilitates the recognition of the variable loop 3 (V3)-glycan patch on the envelope protein of HIV-1. RC1 conceals non-conserved immunodominant regions by the addition of glycans and/or multimerization on virus-like particles. Immunization of mice, rabbits and rhesus macaques with RC1 elicited serological responses that targeted the V3-glycan patch. Antibody cloning and cryo-electron microscopy structures of antibody-envelope complexes confirmed that immunization with RC1 expands clones of B cells that carry the anti-V3-glycan patch antibodies, which resemble precursors of human broadly neutralizing antibodies. Thus, RC1 may be a suitable priming immunogen for sequential vaccination strategies in the context of polyclonal repertoires.