Merck
  • Resorcinol Functionalized Gold Nanoparticles for Formaldehyde Colorimetric Detection.
Resorcinol Functionalized Gold Nanoparticles for Formaldehyde Colorimetric Detection.
Nanomaterials (Basel, Switzerland) (2019-03-01)
Carlos Martínez-Aquino, Ana M Costero, Salvador Gil, Pablo Gaviña
ABSTRACT
Gold nanoparticles functionalized with resorcinol moieties have been prepared and used for detecting formaldehyde both in solution and gas phases. The detection mechanism is based on the color change of the probe upon the aggregation of the nanoparticles induced by the polymerization of the resorcinol moieties in the presence of formaldehyde. A limit of detection of 0.5 ppm in solution has been determined. The probe can be deployed for the detection of formaldehyde emissions from composite wood boards.