Merck
Notch Signaling Mediates Secondary Senescence.
Cell reports (2019-04-25)
Yee Voan Teo, Nattaphong Rattanavirotkul, Nelly Olova, Angela Salzano, Andrea Quintanilla, Nuria Tarrats, Christos Kiourtis, Miryam Müller, Anthony R Green, Peter D Adams, Juan-Carlos Acosta, Thomas G Bird, Kristina Kirschner, Nicola Neretti, Tamir Chandra
ABSTRACT
Oncogene-induced senescence (OIS) is a tumor suppressive response to oncogene activation that can be transmitted to neighboring cells through secreted factors of the senescence-associated secretory phenotype (SASP). Currently, primary and secondary senescent cells are not considered functionally distinct endpoints. Using single-cell analysis, we observed two distinct transcriptional endpoints, a primary endpoint marked by Ras and a secondary endpoint marked by Notch activation. We find that secondary oncogene-induced senescence in vitro and in vivo requires Notch, rather than SASP alone, as previously thought. Moreover, Notch signaling weakens, but does not abolish, SASP in secondary senescence. Global transcriptomic differences, a blunted SASP response, and the induction of fibrillar collagens in secondary senescence point toward a functional diversification between secondary and primary senescence.
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Pricing and availability is not currently available.