Merck
  • Platelet-derived growth factor activates nociceptive neurons by inhibiting M-current and contributes to inflammatory pain.
Platelet-derived growth factor activates nociceptive neurons by inhibiting M-current and contributes to inflammatory pain.
Pain (2019-04-02)
Omer Barkai, Stephanie Puig, Shaya Lev, Ben Title, Ben Katz, Luba Eli-Berchoer, Howard B Gutstein, Alexander M Binshtok
ABSTRACT
Endogenous inflammatory mediators contribute to the pathogenesis of pain by acting on nociceptors, specialized sensory neurons that detect noxious stimuli. Here, we describe a new factor mediating inflammatory pain. We show that platelet-derived growth factor (PDGF)-BB applied in vitro causes repetitive firing of dissociated nociceptor-like rat dorsal root ganglion neurons and decreased their threshold for action potential generation. Injection of PDGF-BB into the paw produced nocifensive behavior in rats and led to thermal and mechanical pain hypersensitivity. We further detailed the biophysical mechanisms of these PDGF-BB effects and show that PDGF receptor-induced inhibition of nociceptive M-current underlies PDGF-BB-mediated nociceptive hyperexcitability. Moreover, in vivo sequestration of PDGF or inhibition of the PDGF receptor attenuates acute formalin-induced inflammatory pain. Our discovery of a new pain-facilitating proinflammatory mediator, which by inhibiting M-current activates nociceptive neurons and thus contributes to inflammatory pain, improves our understanding of inflammatory pain pathophysiology and may have important clinical implications for pain treatment.