Merck
  • Inhibition of microRNA-155 attenuates sympathetic neural remodeling following myocardial infarction via reducing M1 macrophage polarization and inflammatory responses in mice.
Inhibition of microRNA-155 attenuates sympathetic neural remodeling following myocardial infarction via reducing M1 macrophage polarization and inflammatory responses in mice.
European journal of pharmacology (2019-02-06)
Juan Hu, Cong-Xin Huang, Pan-Pan Rao, Ji-Peng Zhou, Xi Wang, Lu Tang, Ming-Xin Liu, Guo-Gang Zhang
ABSTRACT
Inflammation plays an important role in sympathetic neural remodeling induced by myocardial infarction (MI). MiR-155 is a vital regulator of inflammatory responses, and macrophage-secreted miR-155 promotes cardiac fibrosis and hypertrophy. However, whether miR-155 influences MI-induced sympathetic neural remodeling is not clear. Therefore, we examined the role of miR-155 in MI-induced sympathetic neural remodeling and the related mechanisms in both an mouse model and in lipopolysaccharide (LPS)-stimulated bone marrow-derived macrophages (BMDMs). Our data showed that miR-155 expression was significantly enhanced in the myocardial tissues of MI mice compared to sham mice. Also, MI up-regulated the electrophysiological parameters, M1 macrophage polarization, inflammatory responses, and suppressor of cytokine signaling 1 (SOCS1) expression, which coincided with the increased expression of sympathetic nerve remodeling markers(nerve growth factor, tyrosine hydroxylase and growth-associated protein 43). Except for SOCS1, these proteins were attenuated by miR-155 antagomir. In vitro, LPS-stimulation promoted miR-155 expression in BMDMs. Consistent with the in vivo findings, miR-155 antagomir diminished the LPS-induced M1 macrophage polarization, nuclear factor (NF)-κB activation, and the expression of pro-inflammatory factors and nerve growth factor; but it increased the expression of SOCS1. Inversely, miR-155 agomir significantly potentiated LPS-induced pathophysiological effects in BMDMs. MiR-155 agomir-induced effects were reversed by the NF-κB inhibitor. Mechanistically, treatment with siRNA against SOCS1 augmented the aforementioned LPS-mediated activities, which were antagonized by the addition of miR-155 antagomir. In conclusion, miR-155 inhibition downregulated NGF expression via decreasing M1 macrophage polarization and inflammatory responses dependent on the SOCS1/NF-κB pathway, subsequently diminishing MI-induced sympathetic neural remodeling and ventricular arrhythmias (VAs).