Merck
  • Soluble guanylyl cyclase activation by HMR-1766 (ataciguat) in cells exposed to oxidative stress.
Soluble guanylyl cyclase activation by HMR-1766 (ataciguat) in cells exposed to oxidative stress.
American journal of physiology. Heart and circulatory physiology (2008-09-02)
Zongmin Zhou, Anastasia Pyriochou, Anastasia Kotanidou, Georgios Dalkas, Martin van Eickels, Georgios Spyroulias, Charis Roussos, Andreas Papapetropoulos
ABSTRACT
Many vascular diseases are characterized by increased levels of ROS that destroy the biological activity of nitric oxide and limit cGMP formation. In the present study, we investigated the cGMP-forming ability of HMR-1766 in cells exposed to oxidative stress. Pretreatment of smooth muscle cells with H(2)O(2) reduced cGMP production stimulated by sodium nitroprusside (SNP) or BAY 41-2272. However, pretreatment with H(2)O(2) significantly increased HMR-1766 responses. Similar results were obtained with SIN-1, menadione, and rotenone. In addition, HMR-1766 was more effective in stimulating heme-free sGC compared with the wild-type enzyme. Interestingly, in cells expressing heme-free sGC, H(2)O(2) inhibited instead of potentiated HMR-1766 responses, suggesting that the ROS-induced enhancement of cGMP formation was heme dependent. Moreover, using truncated forms of sGC, we observed that the NH(2)-terminus of the beta(1)-subunit is required for the action of HMR-1766. Finally, to study tolerance development to HMR-1766, cells were pretreated with this sGC activator and reexposed to HMR-1766 or SNP. Results from these experiments demonstrated lack of tolerance development to HMR-1766 as well as lack of cross-tolerance with SNP. We conclude that HMR-1766 is an improved sGC activator as it has the ability to activate oxidized/heme-free sGC and is resistant to the development of tolerance; these observations make HMR-1766 a promising agent for treating diseases associated with increased vascular tone combined with enhanced ROS production.
Sigma-Aldrich
Ataciguat, ≥98% (HPLC)
Pricing and availability is not currently available.