Merck
  • A spectrophotometric assay for nanogram quantities of biotin and avidin.
A spectrophotometric assay for nanogram quantities of biotin and avidin.
Journal of biochemical and biophysical methods (1986-11-01)
R S Niedbala, F Gergits, K J Schray
ABSTRACT
Parameters and conditions of an enzyme based assay for biotin and avidin are presented. Biotinylated glucose-6-phosphate dehydrogenase when complexed with avidin becomes inactivated. Thus it was possible to construct a competitive assay system for biotin. The assay is sensitive between 100-500 ng/ml and could detect as little as 10 ng in 0.1 ml with a between run error of 2.4%. It requires a 60 min incubation at 21 degrees C and 5 min to assay. The avidin assay, based on the degree of inactivation of biotinylated-glucose-6-phosphate dehydrogenase in relation to the concentration of avidin, could detect as little as 0.25 ng in 0.1 ml or 2.5 ng/ml with an assay time of 10 min with a between run error of 3.9%. Both assays are rapid with significant improvements over other non-isotopic methods in sensitivity and comparable to radioisotopic methods in sensitivity with the added advantage of ease of method.
Sigma-Aldrich
HABA/Avidin Reagent, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Streptavidin–β-Galactosidase conjugate, streptavidin 1-2 units/mg protein, lyophilized powder
Pricing and availability is not currently available.