Merck
  • Molecular characterization of flavonoid malonyltransferase from Oryza sativa.
Molecular characterization of flavonoid malonyltransferase from Oryza sativa.
Plant physiology and biochemistry : PPB (2009-09-08)
Dea Hwan Kim, Seong Kyong Kim, Jeong-Ho Kim, Bong-Gyu Kim, Joong-Hoon Ahn
ABSTRACT
In this study, a flavonoid malonyltransferase (OsMaT-2) was cloned from Oryza sativa, and the recombinant protein OsMaT-2 was purified via affinity chromatography. OsMaT-2 utilized a variety of flavonoid glucosides, including flavanone glucosides, flavone glucosides, flavonol glucosides, and isoflavone glucosides as substrates, but did not utilize anthocyanin. As an acyl donor, OsMaT-2 utilized only malonyl-CoA. Based on reactions with various quercetin 3-O-sugars, we identified the probable position of malonylation as the 6''-hydroxyl group of the sugar. This is the first report, to the best of our knowledge, of the cloning of a flavonoid malonyltransferase from O. sativa.
Sigma-Aldrich
Malonyl coenzyme A tetralithium salt, ≥90% (HPLC)