- Dealing with energy demand: the AMP-activated protein kinase.
Dealing with energy demand: the AMP-activated protein kinase.
The AMP-activated protein kinase (AMPK) is a member of a metabolite-sensing protein kinase family that is found in all eukaryotes. AMPK activity is regulated by vigorous exercise, nutrient starvation and ischemia/hypoxia, and modulates many aspects of mammalian cell metabolism. The AMPK yeast homolog, Snf1p, plays a major role in adaption to glucose deprivation. In mammals, AMPK also has diverse roles that extend from energy metabolism through to transcriptional control.