Merck
  • Tension-loaded bone marrow stromal cells potentiate the paracrine osteogenic signaling of co-cultured vascular endothelial cells.
Tension-loaded bone marrow stromal cells potentiate the paracrine osteogenic signaling of co-cultured vascular endothelial cells.
Biology open (2018-05-03)
Yu Nan Jiang, Jun Zhao, Feng Ting Chu, Yang Yang Jiang, Guo Hua Tang
ABSTRACT
Co-culture of bone marrow stromal cells (BMSCs) and vascular endothelial cells (VECs) is a promising strategy for better osteogenesis and pre-vascularization in bone tissue engineering. Recent reports have shown that mechanical stretching further promotes osteogenesis in BMSC/VEC co-culture systems, but the underlying mechanism of this process remains unclear. In this study, noncontact co-cultures of rat primary BMSCs and VECs were employed to interrogate paracrine cell-to-cell communications in response to tension. Exposure of VECs to 6% tension for 48 h elicited neither ALP activity nor mRNA expression of OCN and OPN in BMSCs incubated in a shared culture medium. Instead, BMSCs subjected to tension induced robust VEGF release, and its conditioned medium enhanced the proliferation and tubular formation of VECs with a concurrent increase in BMP-2 and IGF-1 production. Conditioned medium from activated VECs in turn promoted expression of osteogenic genes in BMSCs, followed by an increase in matrix mineralization. The addition of VEGF-R inhibitor Tivozanib to these systems abrogated the tension-induced paracrine effects on VECs and subsequently impaired BMSC osteogenesis. These results clearly demonstrate that the response of BMSCs to tension potentiates paracrine osteogenic signaling from VECs; this positive feedback loop is initiated by VEGF release.
Sigma-Aldrich
4-Vinyl-1,3-dioxolan-2-one, 99%
Pricing and availability is not currently available.