Merck
  • Synthesis and application of ionic liquid functionalized β-cyclodextrin, mono-6-deoxy-6-(4-amino-1,2,4-triazolium)-β-cyclodextrin chloride, as chiral selector in capillary electrophoresis.
Synthesis and application of ionic liquid functionalized β-cyclodextrin, mono-6-deoxy-6-(4-amino-1,2,4-triazolium)-β-cyclodextrin chloride, as chiral selector in capillary electrophoresis.
Journal of chromatography. A (2018-01-07)
Jingtang Li, Tao Yu, Guangfu Xu, Yingxiang Du, Zongran Liu, Zijie Feng, Xuan Yang, Ying Xi, Jie Liu
ABSTRACT
Recently,ionic liquids (ILs) functionalized cyclodextrins (CDs) have attracted more and more attention in the fields of enantioseparation. In this study, a novel IL amino triazolium functionalized β-CD derivative, mono-6-deoxy-6-(4-amino-1,2,4-triazolium)-β-cyclodextrin chloride (4-ATMCDCl), was synthesized for the first time and managed to separate dansyl amino acids and naproxen by capillary electrophoresis (CE). Compared with native β-CD, the new selector exhibited good water solubility and enhanced enantioselectivity. Several crucial parameters such as selector concentration, buffer pH, and applied voltage were systematically investigated. The molecular docking program Autodock was applied to further demonstrate the mechanism of chiral recognition and the enhanced enantioselectivity of 4-ATMCDCl, which showed good agreement with our experimental results.
Sigma-Aldrich
4-Amino-4H-1,2,4-triazole, ReagentPlus®, 99%