
Urea Crystals

Synonym(s):
Urea, Carbamide, Carbonyldiamide
Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Products
Formula Weight
Melting Point (°C)
Product Line
Markush Class
Markush Group
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing