Skip to Content
Merck
vibrant-m

Theophylline Related Compound C

vibrant-m
Synonym(s):
N-(6-Amino-1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)formamide, Theophylline Related Compound C
Empirical Formula (Hill Notation):
C7H10N4O3
CAS Number:
Molecular Weight:
198.18

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1