Skip to Content
ZA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products/content
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethanol
Synonym(s):
Ethanol
Empirical Formula (Hill Notation):
C
2
H
6
O
Molecular Weight:
46.07
Products
Formula Weight
Physical Form
Product Line
Format
Package Size
Special Grade
Absolute (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SAAR2233510
Ethanol,
rectified, 96%, univAR
™
Expand
View Pricing
100983L
Ethanol,
absolute, GR For Analysis
Expand
View Pricing