
Eclalbasaponin II

Synonym(s):
(3β,16α)-3-(β-D-Glucopyranosyloxy)-16-hydroxyolean-12-en-28-oic acid, Ecliptasaponin A
Empirical Formula (Hill Notation):
C36H58O9
CAS Number:
Molecular Weight:
634.84
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing