Charcoal
Synonym(s):
Activated charcoal, Charcoal activated
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Products
Formula Weight
Melting Point (°C)
Physical Form
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing