Skip to Content
Merck
vibrant-m

Cefepime dihydrochloride monohydrate

vibrant-m
Empirical Formula (Hill Notation):
C19H25ClN6O5S2 · HCl · H2O
CAS Number:
Molecular Weight:
571.50

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1